Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081756.1 | NP_001075225.1 | 7 | 925 | |
NM_172484.3 | NP_766072.2 | 5 | 786 | ||
NM_176957.3 | NP_795931.2 | 7 | 925 | ||
GenBank mRNA | AK053243.1 | - | 5 | 608 | |
AY957601.1 | - | 3 | 345 | ||
AY957602.1 | - | 3 | 345 | ||
BC066095.1 | - | 5 | 786 | 6856184 | |
BC079551.1 | - | 5 | 786 | 6856184 |