Gene Transcripts

Gene Symbol
Plk5
Entrez Gene ID
216166
Gene Name
polo-like kinase 5
Gene Aliases
6330514A18Rik
Chromosome Location
Chr. 10: 80356459 - 80365489 on Build GRCm38
UniGene ID
Mm.17613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183152.3 NP_898975.2 4, 5 578
GenBank mRNA AK031968.1 - 4, 5 580
BB622163.1 - 4, 5 579
BC099679.1 - 4, 5 547 6489647
BG297060.1 - 2, 3 165

View Full Product Details