Gene Transcripts

Gene Symbol
Gramd4
Entrez Gene ID
223752
Gene Name
GRAM domain containing 4
Gene Aliases
9930016O13
Chromosome Location
Chr. 15: 86057695 - 86137636 on Build GRCm38
UniGene ID
Mm.24442
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205353.1 NP_001192282.1 18 2000
NM_172611.4 NP_766199.1 18 1807
GenBank mRNA AK036832.1 - 18 1796
AK129209.1 - 14 3782
AK142266.1 - 17 2556
BC021523.1 - 14 1181
BC085148.1 - 18 1958
BC139228.1 - 18 1796 8862250
BC139229.1 - 18 1796 8862251

View Full Product Details