Gene Transcripts

Gene Symbol
Sec16a
Entrez Gene ID
227648
Gene Name
SEC16 homolog A (S. cerevisiae)
Gene Aliases
AU024582, C230052J16Rik
Chromosome Location
Chr. 2: 26409431 - 26446162 on Build GRCm38
UniGene ID
Mm.206371
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153125.2 NP_694765.2 11, 12 5159
GenBank mRNA AK122242.2 - 4, 5 596
BC034649.1 - 1, 2 217
BC042603.1 - 5, 6 675
BC059898.1 - 2, 3 763

View Full Product Details