Gene Transcripts

Gene Symbol
Pdap1
Entrez Gene ID
231887
Gene Name
PDGFA associated protein 1
Gene Aliases
HASPP28, PAP, PAP1
Chromosome Location
Chr. 5: 145128770 - 145140089 on Build GRCm38
UniGene ID
Mm.188851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033313.3 NP_001028485.1 5 482
GenBank mRNA AK147170.1 - 5 483
AK161518.1 - 4 611
AK169233.1 - 5 416
BC007162.1 - 4 382
BC055927.1 - 5 396
BC106168.1 - 4 370
BC114598.1 - 3 216
BC139204.1 - 5 435 8862226
BC139205.1 - 5 435 8862227

View Full Product Details