Gene Transcripts

Gene Symbol
Sc5d
Entrez Gene ID
235293
Gene Name
sterol-C5-desaturase (fungal ERG3, delta-5-desaturase) homolog (S. cerevisae)
Gene Aliases
A830037K02, A830073K23Rik, Sc5dl
Chromosome Location
Chr. 9: 42251592 - 42264300 on Build GRCm38
UniGene ID
Mm.32700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172769.2 NP_766357.1 6 1064
GenBank mRNA AK043825.1 - 6 1023
AK077670.1 - 6 1022

View Full Product Details