Gene Transcripts

Gene Symbol
Macc1
Entrez Gene ID
238455
Gene Name
metastasis associated in colon cancer 1
Gene Aliases
4732474O15Rik, Gm267
Chromosome Location
Chr. 12: 119326285 - 119466934 on Build GRCm38
UniGene ID
Mm.31376
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163136.1 NP_001156608.1 2 1914
GenBank mRNA AK028965.1 - 1 236

View Full Product Details