Gene Transcripts

Gene Symbol
Emilin2
Entrez Gene ID
246707
Gene Name
elastin microfibril interfacer 2
Gene Aliases
FOAP-10
Chromosome Location
Chr. 17: 71252172 - 71311639 on Build GRCm38
UniGene ID
Mm.23462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145158.3 NP_660140.1 4 2303
GenBank mRNA AF468645.1 - 4 2254
AK154230.1 - 4 2273
AK155037.1 - 4 2283
AK155306.1 - 4 2297
AK155366.1 - 4 2283
AK155554.1 - 4 2299
AK155797.1 - 4 2304
AK155878.1 - 4 2301
AK155879.1 - 4 2297
AK155909.1 - 4 2303
AK155913.1 - 4 2273
AK155918.1 - 4 2330
AK170081.1 - 4 2300
AK170288.1 - 4 2273
AK170391.1 - 4 2273
AK170801.1 - 4 2304
AK170822.1 - 4 2325
AK170953.1 - 4 2319
AK171015.1 - 4 2325
AK171020.1 - 4 2291
AK171303.1 - 4 2275
AK171309.1 - 4 2300
AK171579.1 - 4 2273
AK171592.1 - 4 2273
AK171616.1 - 4 2273
BC053753.1 - 4 2587 6510220
GQ872457.1 - 4 2198
GQ872460.1 - 4 2198

View Full Product Details