Gene Transcripts

Gene Symbol
Olfr464
Entrez Gene ID
258407
Gene Name
olfactory receptor 464
Gene Aliases
MOR240-2
Chromosome Location
Chr. 11: 87913920 - 87915003 on Build GRCm38
UniGene ID
Mm.271337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146412.2 NP_666524.2 1 267
GenBank mRNA BC120701.1 - 1 268

View Full Product Details