Gene Transcripts
- Gene Symbol
- Nbea
- Entrez Gene ID
- 26422
- Gene Name
- neurobeachin
- Gene Aliases
- Lyst2, mKIAA1544
- Chromosome Location
- Chr. 3: 55625195 - 56183701 on Build GRCm38
- UniGene ID
- Mm.384353
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030595.1 |
NP_085098.1
|
18 |
2830 |
|
GenBank mRNA |
Y18276.1 |
- |
18 |
2830 |
|
View Full Product Details