Gene Transcripts

Gene Symbol
Slc27a3
Entrez Gene ID
26568
Gene Name
solute carrier family 27 (fatty acid transporter), member 3
Gene Aliases
Acsvl3, FATP3, Vlcs-3
Chromosome Location
Chr. 3: 90385228 - 90390042 on Build GRCm38
UniGene ID
Mm.45519
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011988.2 NP_036118.2 5 1387
GenBank mRNA AF072758.1 - 5 1091
AJ577573.1 - 5 1387
AK037042.1 - 5 1147
AK076014.1 - 5 1119
AK131156.1 - 3 2164
AK135190.1 - 5 1364
BC060052.1 - 5 1395
BC141134.1 - 5 1352 9055746

View Full Product Details