Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012002.3 | NP_036132.1 | 9 | 771 | |
GenBank mRNA | AF071315.1 | - | 9 | 808 | |
AK138409.1 | - | 7 | 986 | ||
AK146410.1 | - | 9 | 782 | ||
AK146775.1 | - | 9 | 784 | ||
BC004664.1 | - | 9 | 770 | 3498214 | |
BC014286.1 | - | 9 | 775 | 4163277 |