Gene Transcripts

Gene Symbol
Cd200r2
Entrez Gene ID
271375
Gene Name
Cd200 receptor 2
Gene Aliases
AY230198, Cd200r1l
Chromosome Location
Chr. 16: 44867097 - 44915840 on Build GRCm38
UniGene ID
Mm.441535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206535.1 NP_996258.1 3 321
GenBank mRNA AY230198.1 - 3 321

View Full Product Details