Gene Transcripts

Gene Symbol
Golga5
Entrez Gene ID
27277
Gene Name
golgi autoantigen, golgin subfamily a, 5
Gene Aliases
Ret-II
Chromosome Location
Chr. 12: 102469134 - 102497911 on Build GRCm38
UniGene ID
Mm.273370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199004.1 NP_001185933.1 2 617
NM_013747.4 NP_038775.1 2 459
GenBank mRNA AB016784.1 - 2 618
AF026274.1 - 2 449
AK007592.1 - 2 464
AK138455.1 - 2 618
AK152533.1 - 2 467
AK153010.1 - 2 467
AK168765.1 - 2 463
BC016883.1 - 2 1187 4235952
BC086782.1 - 2 459 6813878

View Full Product Details