Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177157.4 | NP_796131.1 | 2 | 111 | |
GenBank mRNA | AK028116.1 | - | 2 | 112 | |
AK028122.1 | - | 2 | 112 | ||
BC096406.1 | - | 2 | 99 | 1511037 | |
BC132450.1 | - | 2 | 107 | 40130727 | |
BC132478.1 | - | 2 | 107 | 40130755 |