Gene Transcripts

Gene Symbol
Usp24
Entrez Gene ID
329908
Gene Name
ubiquitin specific peptidase 24
Gene Aliases
2700066K03Rik, 2810030C21Rik, AI414051, B130021E18, C79851
Chromosome Location
Chr. 4: 106315899 - 106446572 on Build GRCm38
UniGene ID
Mm.234544
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183225.2 NP_899048.2 45 5574

View Full Product Details