Gene Transcripts

Gene Symbol
Ctxn1
Entrez Gene ID
330695
Gene Name
cortexin 1
Gene Aliases
BC028881, Ctxn
Chromosome Location
Chr. 8: 4257646 - 4259274 on Build GRCm38
UniGene ID
Mm.34210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183315.2 NP_899138.1 2 362
GenBank mRNA AK154120.1 - 2 314
AK158759.1 - 2 256
BC028881.1 - 2 298 4224851

View Full Product Details