Gene Transcripts

Gene Symbol
Zscan10
Entrez Gene ID
332221
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
BC068284, Zfp206, Zkscan10, Znf206
Chromosome Location
Chr. 17: 23599751 - 23611019 on Build GRCm38
UniGene ID
Mm.270315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033425.4 NP_001028597.2 6 1172
NM_001289482.1 NP_001276411.1 4 842
GenBank mRNA AK141259.1 - 6 1143
BC068284.1 - 5 845
DQ323929.1 - 6 1140
DQ323931.1 - 4 810
EF152498.1 - 5 987
EF152502.1 - 4 764
EF152503.1 - 2 641

View Full Product Details