Gene Transcripts
- Gene Symbol
- E030044B06Rik
- Entrez Gene ID
- 381223
- Gene Name
- RIKEN cDNA E030044B06 gene
- Gene Aliases
- E030007J07Rik, Gm1618
- Chromosome Location
- Chr. 19: 40868316 - 40882639 on Build GRCm38
- UniGene ID
- Mm.136844
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087313.1 |
- |
3 |
253 |
|
View Full Product Details