Gene Transcripts

Gene Symbol
Irs2
Entrez Gene ID
384783
Gene Name
insulin receptor substrate 2
Gene Aliases
Irs-2
Chromosome Location
Chr. 8: 10986961 - 11008430 on Build GRCm38
UniGene ID
Mm.407207
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081212.1 NP_001074681.1 1 3282
GenBank mRNA AK155277.1 - 1 3104
BC147580.1 - 1 3310 9056570

View Full Product Details