Gene Transcripts

Gene Symbol
Ccdc120
Entrez Gene ID
54648
Gene Name
coiled-coil domain containing 120
Gene Aliases
AA030924, DXImx50e, Jm11, Sfc25
Chromosome Location
Chr. X: 7731714 - 7741324 on Build GRCm38
UniGene ID
Mm.63342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207202.2 NP_997085.2 6 679
GenBank mRNA BC053918.1 - 6 642
BC139064.1 - 5 455 8862086

View Full Product Details