Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019685.2 | NP_062659.1 | 2 | 258 | |
GenBank mRNA | AF100694.1 | - | 2 | 297 | |
AK012170.1 | - | 2 | 206 | ||
AK132783.1 | - | 2 | 240 | ||
AK145062.1 | - | 207 | |||
AK146258.1 | - | 2 | 250 | ||
AK146374.1 | - | 2 | 251 | ||
AK156078.1 | - | 2 | 228 | ||
AK183435.1 | - | 2 | 50 | ||
AK188570.1 | - | 2 | 50 | ||
BC004718.1 | - | 2 | 224 | 3582426 | |
CJ146900.1 | - | 2 | 260 |