Gene Transcripts

Gene Symbol
Chchd1
Entrez Gene ID
66121
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 1
Gene Aliases
1110001O19Rik, 2400010G13Rik, MRP-S37
Chromosome Location
Chr. 14: 20703027 - 20704425 on Build GRCm38
UniGene ID
Mm.246996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025366.3 NP_079642.1 3 386
GenBank mRNA AK003263.1 - 3 345
AK010311.1 - 3 329
AK011261.1 - 3 344
AK027907.1 - 3 370
AV460497.1 - 3 386
BC049582.1 - 369 6772118
BC052720.1 - 3 365 6818266
BC064310.1 - 3 365
BC092216.1 - 3 353 30609530

View Full Product Details