Gene Transcripts

Gene Symbol
Prr13
Entrez Gene ID
66151
Gene Name
proline rich 13
Gene Aliases
1110020C13Rik, 2010324E22Rik
Chromosome Location
Chr. 15: 102459170 - 102462806 on Build GRCm38
UniGene ID
Mm.393955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170911.1 NP_001164382.1 3, 4 471
NM_025385.3 NP_079661.1 4, 5 589
GenBank mRNA AK002430.1 - 3, 4 439
AK003850.1 - 4, 5 586
AK008611.1 - 2, 3 411
AK008894.1 - 4, 5 549
AK154583.1 - 4, 5 546
AK181565.1 - 1, 2 246
AK218697.1 - 1, 2 139
AK219155.1 - 1, 2 207
BC016234.1 - 3, 4 735 4503179

View Full Product Details