Gene Transcripts

Gene Symbol
Topaz1
Entrez Gene ID
671232
Gene Name
testis and ovary specific PAZ domain containing 1
Gene Aliases
Gm9524
Chromosome Location
Chr. 9: 122747217 - 122802074 on Build GRCm38
UniGene ID
Mm.296421
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199736.1 NP_001186665.1 16 4107
GenBank mRNA HM631980.1 - 16 4107

View Full Product Details