Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025988.2 | NP_080264.1 | 8 | 1207 | |
GenBank mRNA | AK008207.1 | - | 2 | 269 | |
AK008243.1 | - | 6 | 756 | ||
AK079604.1 | - | 7 | 1076 | ||
AK148438.1 | - | 702 | |||
AK165381.1 | - | 2 | 344 | ||
BC048371.1 | - | 7 | 1121 | 6396076 |