Gene Transcripts

Gene Symbol
Cwc25
Entrez Gene ID
67480
Gene Name
CWC25 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
1300013D05Rik, AA473636, Ccdc49, R75228
Chromosome Location
Chr. 11: 97745470 - 97766613 on Build GRCm38
UniGene ID
Mm.33206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026186.4 NP_080462.2 3 584
GenBank mRNA AK004983.2 - 3 528
AK017849.1 - 3 529
AK030628.1 - 3 513
AK035158.1 - 3 513
BC021478.1 - 3 498
BC079553.1 - 3 506 6856335

View Full Product Details