Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026348.3 | NP_080624.1 | 3, 4 | 203 | |
GenBank mRNA | AK005779.1 | - | 3, 4 | 272 | |
AK013373.1 | - | 3, 4 | 218 | ||
AK015557.1 | - | 3, 4 | 228 | ||
AK045037.1 | - | 3, 4 | 204 | ||
AK166362.1 | - | 3, 4 | 204 | ||
BC049557.1 | - | 3, 4 | 213 | 6741650 | |
EF437329.1 | - | 1, 2 | 111 |