Gene Transcripts

Gene Symbol
Fam96a
Entrez Gene ID
68250
Gene Name
family with sequence similarity 96, member A
Gene Aliases
5730536A07Rik
Chromosome Location
Chr. 9: 66126611 - 66138968 on Build GRCm38
UniGene ID
Mm.27107
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026635.3 NP_080911.1 4, 5 623
GenBank mRNA AK002690.1 - 4, 5 565
AK003870.1 - 4, 5 519
AK077700.1 - 4, 5 624
AK170445.1 - 4, 5 417
AK179061.1 - 3, 4 143
AK184756.1 - 2, 3 72
BC005745.1 - 4, 5 540 3600028

View Full Product Details