Gene Transcripts

Gene Symbol
Lypd2
Entrez Gene ID
68311
Gene Name
Ly6/Plaur domain containing 2
Gene Aliases
0610005K03Rik, Lypdc2, VLL
Chromosome Location
Chr. 15: 74732246 - 74734795 on Build GRCm38
UniGene ID
Mm.306880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026671.1 NP_080947.1 1, 2 94
GenBank mRNA AB009019.1 - 1, 2 110
AK002226.1 - 1, 2 110
BC099511.1 - 1, 2 94 30845404
BC132407.1 - 1, 2 95 40130684
BC132409.1 - 1, 2 95 40130686

View Full Product Details