Gene Transcripts

Gene Symbol
Chac1
Entrez Gene ID
69065
Gene Name
ChaC, cation transport regulator 1
Gene Aliases
1810008K03Rik
Chromosome Location
Chr. 2: 119351242 - 119354326 on Build GRCm38
UniGene ID
Mm.35083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026929.4 NP_081205.1 1 290
GenBank mRNA AK007378.1 - 1 243
AK032218.1 - 1 276
BC025169.1 - 1 231 4483043

View Full Product Details