Gene Transcripts

Gene Symbol
Stoml1
Entrez Gene ID
69106
Gene Name
stomatin-like 1
Gene Aliases
1810015E19Rik, AI847431, SLP1, STORP, UNC24, WPB72
Chromosome Location
Chr. 9: 58253164 - 58262524 on Build GRCm38
UniGene ID
Mm.216502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026942.3 NP_081218.3 2 328
GenBank mRNA AK045820.1 - 2 300
AK148324.1 - 227
BC017536.1 - 2 300
BC037074.1 - 2 258 5037636

View Full Product Details