Gene Transcripts
- Gene Symbol
- Glipr1l1
- Entrez Gene ID
- 69286
- Gene Name
- GLI pathogenesis-related 1 like 1
- Gene Aliases
- 1700011E04Rik
- Chromosome Location
- Chr. 10: 112060189 - 112078510 on Build GRCm38
- UniGene ID
- Mm.47058
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027018.1 |
NP_081294.1
|
3, 4 |
667 |
|
GenBank mRNA |
AK005860.1 |
- |
3, 4 |
668 |
|
View Full Product Details