Gene Transcripts

Gene Symbol
Tmem52
Entrez Gene ID
69671
Gene Name
transmembrane protein 52
Gene Aliases
2310043I08Rik
Chromosome Location
Chr. 4: 155469114 - 155470858 on Build GRCm38
UniGene ID
Mm.45401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253856.1 NP_001240785.1 4, 5 373
NM_001253858.1 NP_001240787.1 3, 4 331
NM_027161.2 NP_081437.1 4, 5 391
GenBank mRNA AK009779.1 - 4, 5 354
BC144992.1 - 4, 5 390 9053540
BC150685.1 - 4, 5 371 9087596

View Full Product Details