Gene Transcripts

Gene Symbol
Unc13d
Entrez Gene ID
70450
Gene Name
unc-13 homolog D (C. elegans)
Gene Aliases
2610108D09Rik, Jinx, Munc13-4, mFLJ00067
Chromosome Location
Chr. 11: 116062095 - 116078195 on Build GRCm38
UniGene ID
Mm.259460
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009573.2 NP_001009573.2 4 340
GenBank mRNA BC046519.1 - 4 340
BC141289.1 - 3 252 9055901
EF127645.1 - 3 288

View Full Product Details