Gene Transcripts

Gene Symbol
Lrrc56
Entrez Gene ID
70552
Gene Name
leucine rich repeat containing 56
Gene Aliases
5730427C23Rik, BB110509, mFLJ00101
Chromosome Location
Chr. 7: 141194116 - 141210055 on Build GRCm38
UniGene ID
Mm.45104
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172064.1 NP_001165535.1 9 1485
NM_001172065.1 NP_001165536.1 9 1485
NM_153777.2 NP_722472.1 9 1071
GenBank mRNA AK017603.1 - 9 1459
AK220237.1 - 9 1439
BC027807.1 - 9 1081 5324022

View Full Product Details