Gene Transcripts

Gene Symbol
Gpr173
Entrez Gene ID
70771
Gene Name
G-protein coupled receptor 173
Gene Aliases
3230401K02Rik, Sreb3
Chromosome Location
Chr. X: 152343598 - 152368968 on Build GRCm38
UniGene ID
Mm.158445
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027543.3 NP_081819.2 3 948
GenBank mRNA AY255602.1 - 1 534
BC043021.1 - 1 716 5686241
BC096520.1 - 3 948 5720034

View Full Product Details