Gene Transcripts

Gene Symbol
Dmrtc1a
Entrez Gene ID
70887
Gene Name
DMRT-like family C1a
Gene Aliases
1700030H14Rik, 4921520P21Rik, DMRTC1, Dmrt8.1
Chromosome Location
Chr. X: 102903216 - 102934003 on Build GRCm38
UniGene ID
Mm.29672
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038616.2 NP_001033705.2 3 350
NM_027591.3 NP_081867.1 5 448
NM_029378.2 NP_083654.1 3 262
GenBank mRNA AK006548.1 - 3 365
AK014934.1 - 5 449
BC147175.1 - 5 420 9056223
BC147176.1 - 5 420 9056216
DQ316772.1 - 5 404
DQ316773.1 - 3 262
DQ316774.1 - 3 262

View Full Product Details