Gene Transcripts

Gene Symbol
Tepp
Entrez Gene ID
73407
Gene Name
testis, prostate and placenta expressed
Gene Aliases
1700055M20Rik
Chromosome Location
Chr. 8: 95311570 - 95326321 on Build GRCm38
UniGene ID
Mm.472783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028532.3 NP_082808.1 3 268
NM_199455.2 NP_955534.1 4 493
GenBank mRNA AK006802.1 - 3 255
BC087870.1 - 4 489 6707505
BC104382.1 - 3 267
BC104383.1 - 3 268
BY706785.1 - 3 253

View Full Product Details