Gene Transcripts
- Gene Symbol
- 4930429H19Rik
- Entrez Gene ID
- 73897
- Gene Name
- RIKEN cDNA 4930429H19 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.387218
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015238.1 |
- |
|
145 |
|
- Gene Symbol
- LOC100505025
- Entrez Gene ID
- 100505025
- Gene Name
- uncharacterized LOC100505025
- Gene Aliases
- Chromosome Location
- Chr. 7: 73310380 - 73315193 on Build GRCm38
- UniGene ID
- Mm.445959
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK133139.1 |
- |
1 |
295 |
|
View Full Product Details