Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029152.1 | NP_083428.1 | 3, 4 | 587 | |
GenBank mRNA | AK015697.1 | - | 3, 4 | 589 | |
BC099564.1 | - | 3, 4 | 587 | ||
BC119387.1 | - | 3, 4 | 482 | ||
BC120662.1 | - | 3, 4 | 482 | ||
BF459438.1 | - | 1, 2 | 103 |