Gene Transcripts

Gene Symbol
Dynlrb2
Entrez Gene ID
75465
Gene Name
dynein light chain roadblock-type 2
Gene Aliases
1700009A04Rik, DNLC2B, Dncl2b
Chromosome Location
Chr. 8: 116505015 - 116515915 on Build GRCm38
UniGene ID
Mm.23114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029297.1 NP_083573.1 2 96
GenBank mRNA AK005789.1 - 2 96
BC048623.1 - 2 137 6743790

View Full Product Details