Gene Transcripts

Gene Symbol
Fam71e1
Entrez Gene ID
75538
Gene Name
family with sequence similarity 71, member E1
Gene Aliases
0610007G24Rik, 1700021N13Rik, 1700021P22Rik, GARI-L5
Chromosome Location
Chr. 7: 44496584 - 44501681 on Build GRCm38
UniGene ID
Mm.258841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028169.1 NP_082445.1 2 230
GenBank mRNA AB473729.1 - 2 130
AK006222.1 - 1 330
AK006229.1 - 1 348
AK018715.1 - 2 224
BC049752.1 - 2 147 6773108
BC100470.1 - 2 150
BC130017.1 - 2 173 40140477

View Full Product Details