Gene Transcripts

Gene Symbol
Fam71e1
Entrez Gene ID
75538
Gene Name
family with sequence similarity 71, member E1
Gene Aliases
0610007G24Rik, 1700021N13Rik, 1700021P22Rik, GARI-L5
Chromosome Location
Chr. 7: 44496584 - 44501681 on Build GRCm38
UniGene ID
Mm.258841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028169.1 NP_082445.1 2, 3 260
GenBank mRNA AB473729.1 - 2, 3 160
AK006229.1 - 1, 2 378
BC130017.1 - 2, 3 203 40140477

View Full Product Details