Gene Transcripts
- Gene Symbol
- Zim2
- Entrez Gene ID
- 76637
- Gene Name
- zinc finger, imprinted 2
- Gene Aliases
- 1700116N21Rik
- Chromosome Location
- Chr. 7: 6651130 - 6658385 on Build GRCm38
- UniGene ID
- Mm.159751
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF401983.1 |
- |
|
136 |
|
View Full Product Details