Gene Transcripts

Gene Symbol
Fam49a
Entrez Gene ID
76820
Gene Name
family with sequence similarity 49, member A
Gene Aliases
2410157M17Rik, 9630047E15, D12Ertd553e
Chromosome Location
Chr. 12: 12262129 - 12379058 on Build GRCm38
UniGene ID
Mm.440802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146119.1 NP_001139591.1 7 738
NM_029758.4 NP_084034.2 8 811
GenBank mRNA AK010821.1 - 6 674
AK047142.1 - 7 701
BC049664.1 - 8 773 6532813
BC052465.1 - 7 719 6834768

View Full Product Details