Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030017.3 | NP_084293.1 | 3 | 422 | |
GenBank mRNA | AK020927.1 | - | 3 | 419 | |
AK039233.1 | - | 3 | 423 | ||
AK164157.1 | - | 3 | 424 | ||
BC016204.1 | - | 3 | 405 | 4527851 | |
BC067213.1 | - | 3 | 389 |