Gene Transcripts

Gene Symbol
Cox7b2
Entrez Gene ID
78174
Gene Name
cytochrome c oxidase subunit VIIb2
Gene Aliases
4930503B16Rik
Chromosome Location
Chr. 5: 71442823 - 71548205 on Build GRCm38
UniGene ID
Mm.60690
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030052.3 NP_084328.1 3 319
GenBank mRNA AK019665.1 - 3 320
AK076833.1 - 3 320
BC059709.1 - 3 314 6741029
BY099104.1 - 3 319

View Full Product Details