Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019573.3 | NP_062519.2 | 8, 9 | 1183 | |
GenBank mRNA | AF187014.1 | - | 8, 9 | 1134 | |
AK050541.1 | - | 8, 9 | 1135 | ||
AK078528.1 | - | 8, 9 | 1136 | ||
BC014716.1 | - | 8, 9 | 1139 | 4241066 | |
EF410109.1 | - | 3, 4 | 801 |