Gene Transcripts

Gene Symbol
Cblc
Entrez Gene ID
80794
Gene Name
Casitas B-lineage lymphoma c
Gene Aliases
2310076I21Rik, 2310079L19Rik, Cbl3
Chromosome Location
Chr. 7: 19779718 - 19796809 on Build GRCm38
UniGene ID
Mm.81698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161844.1 NP_001155316.1 1 192
NM_023224.5 NP_075713.3 1 192
GenBank mRNA AF319956.1 - 1 165
AK009399.1 - 1 191
AK010200.1 - 1 190
AK010228.1 - 1 193
BC046337.1 - 1 168 6395806
BC111037.1 - 1 176 5353413

View Full Product Details